Sudan IV
Sudan IV
Sudan IV
Thermo Scientific Chemicals

Sudan IV

CAS: 85-83-6 | C24H20N4O | 380.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A12181.22
also known as A12181-22
Price (EUR)
65,80
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
65,80
Each
Chemical Identifiers
CAS85-83-6
IUPAC Name(1Z)-1-(2-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-1,2-dihydronaphthalen-2-one
Molecular FormulaC24H20N4O
InChI KeyKMDLOETUWUPGMB-BXCCFQQFSA-N
SMILESCC1=CC(=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12)N=NC1=CC=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red to purple or brown to dark brown
FormPowder
Assay (Dye)95.0-105.0%
Loss on Drying<2.5% (105°C/constant wt.)
lambda-max (UV-Vis)514-518 nm (in methanol)
Useful as a stain for fat in animal tissue

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only