DL-Alanine, 99%
DL-Alanine, 99%
DL-Alanine, 99%
Thermo Scientific Chemicals

DL-Alanine, 99%

CAS: 302-72-7 | C3H7NO2 | 89.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A12230.0B
also known as A12230-0B
Price (EUR)
109,65
Online exclusive
122,00
Save 12,35 (10%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (EUR)
109,65
Online exclusive
122,00
Save 12,35 (10%)
Each
Chemical Identifiers
CAS302-72-7
IUPAC Name2-aminopropanoic acid
Molecular FormulaC3H7NO2
InChI KeyQNAYBMKLOCPYGJ-UHFFFAOYNA-N
SMILESCC(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms
View more
DL-Alanine is used in mixed ligand (chelate) studies with transition metals such as Cu(II), Zn(ll) and Cd(ll). It is a functional component in the alanine cycle which regulates the level of alanine aminotransferases which can be used as a diagnostic tool to predict type II diabetes. It is used as a model reagent for development of amino acid racemic resolution techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Alanine is used in mixed ligand (chelate) studies with transition metals such as Cu(II), Zn(ll) and Cd(ll). It is a functional component in the alanine cycle which regulates the level of alanine aminotransferases which can be used as a diagnostic tool to predict type II diabetes. It is used as a model reagent for development of amino acid racemic resolution techniques.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. El-Gamel NE. Uranyl binary and ternary chelates of tenoxicam Synthesis, spectroscopic and thermal characterization of ternary chelates of tenoxicam and alanine with transition metals. Acta A. 2007, 68 860-866.
  2. A H Amrallah; N A Abdalla; E Y El-Haty; Ahmed H Amrallah; Nadia A Abdalla; Esam Y El-Haty. Mixed ligand complexes of benzimidazole and pyrimidine hydroxy azo dyes with some transition metals and glycine, dl-alanine or dl-leucine. Talanta. 1998, 46 (4), 491-500.