Pyridine-2,6-dicarboxylic acid, 98%
Pyridine-2,6-dicarboxylic acid, 98%
Pyridine-2,6-dicarboxylic acid, 98%
Thermo Scientific Chemicals

Pyridine-2,6-dicarboxylic acid, 98%

CAS: 499-83-2 | C7H5NO4 | 167.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A12263.36
also known as A12263-36
Price (EUR)
239,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
239,00
Each
Chemical Identifiers
CAS499-83-2
IUPAC Namepyridine-2,6-dicarboxylic acid
Molecular FormulaC7H5NO4
InChI KeyWJJMNDUMQPNECX-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(=N1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5% (Non-U.S. specification)
Assay (Silylated GC)≥97.5% (Non-U.S. Specification)
Assay from Suppliers CofA≥97.5% (HPLC, U.S. specification)
View more
Pyridine-2,6-dicarboxylic acid is used in the preparation of dipicolinato ligated lanthanide and transition metal complexes. It acts as a chelating agent for chromium, zinc, manganese, copper, iron and molybdenum. Its calcium-dipcolinic acid complex is used to protect deoxyribonucleic acid (DNA) from heat denaturation which enhances the DNA stability. It plays an important role as a marker for the effectiveness of sterilization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine-2,6-dicarboxylic acid is used in the preparation of dipicolinato ligated lanthanide and transition metal complexes. It acts as a chelating agent for chromium, zinc, manganese, copper, iron and molybdenum. Its calcium-dipcolinic acid complex is used to protect deoxyribonucleic acid (DNA) from heat denaturation which enhances the DNA stability. It plays an important role as a marker for the effectiveness of sterilization.

Solubility
Insoluble in water.

Notes
Incompatible with acids, bases and oxidizing agents.
RUO – Research Use Only

General References:

  1. Ge, J.; Yang, Q.; Xie, G.; Chen, S.; Gao, S. Synthesis, structure and thermochemical study of a cobalt energetic coordination compound incorporating 3, 5-diamino-1, 2, 4-triazole and pyridine-2, 6-dicarboxylic acid. J. Chem. Thermodyn. 2015, 80, 1-6.
  2. Mohammadi, S.; Khayatian, G. Colorimetric detection of Bi(III) in water and drug samples using pyridine-2, 6-dicarboxylic acid modified silver nanoparticles. Spectrochim. Acta, Part A 2015, 148, 405-411.