4-Ethoxybenzoic acid, 98+%
4-Ethoxybenzoic acid, 98+%
4-Ethoxybenzoic acid, 98+%
Thermo Scientific Chemicals

4-Ethoxybenzoic acid, 98+%

CAS: 619-86-3 | C9H10O3 | 166.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A12309.22
also known as A12309-22
Price (EUR)
106,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
106,00
Each
Chemical Identifiers
CAS619-86-3
IUPAC Name4-ethoxybenzoic acid
Molecular FormulaC9H10O3
InChI KeySHSGDXCJYVZFTP-UHFFFAOYSA-N
SMILESCCOC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (Silylated GC)≥98.0%
Melting Point (clear melt)195.0-201.0°C
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only