2,5-Dimethylpyrazine, 99%
2,5-Dimethylpyrazine, 99%
2,5-Dimethylpyrazine, 99%
Thermo Scientific Chemicals

2,5-Dimethylpyrazine, 99%

CAS: 123-32-0 | C6H8N2 | 108.144 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12352.06
also known as A12352-06
Price (EUR)
50,10
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
50,10
Each
Chemical Identifiers
CAS123-32-0
IUPAC Name2,5-dimethylpyrazine
Molecular FormulaC6H8N2
InChI KeyLCZUOKDVTBMCMX-UHFFFAOYSA-N
SMILESCC1=CN=C(C)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Identification (FTIR)Conforms
Refractive Index1.4990-1.5020 @ 20?C
FormLiquid
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only