Hydantoin, 99%
Hydantoin, 99%
Hydantoin, 99%
Thermo Scientific Chemicals

Hydantoin, 99%

CAS: 461-72-3 | C3H4N2O2 | 100.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A12486.0E
also known as A12486-0E
Price (EUR)
384,00
Each
Quantity:
2500 g
Request bulk or custom format
Price (EUR)
384,00
Each
Chemical Identifiers
CAS461-72-3
IUPAC Nameimidazolidine-2,4-dione
Molecular FormulaC3H4N2O2
InChI KeyWJRBRSLFGCUECM-UHFFFAOYSA-N
SMILESO=C1CNC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Appearance (Color)White to cream or pale yellow
Assay (HPLC)≥98.5%
Melting Point (clear melt)220-227?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only