Thiophene-2-carboxylic acid, 99%
Thiophene-2-carboxylic acid, 99%
Thiophene-2-carboxylic acid, 99%
Thiophene-2-carboxylic acid, 99%
Thermo Scientific Chemicals

Thiophene-2-carboxylic acid, 99%

CAS: 527-72-0 | C5H4O2S | 128.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A12514.14
also known as A12514-14
Price (EUR)
22,36
Online Exclusive
26,30
Save 3,94 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
22,36
Online Exclusive
26,30
Save 3,94 (15%)
Each
Add to cart
Chemical Identifiers
CAS527-72-0
IUPAC Namethiophene-2-carboxylic acid
Molecular FormulaC5H4O2S
InChI KeyQERYCTSHXKAMIS-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)126-132?C
Assay (GC)≥98.5%
A hypoglycemic, anti-lipolytic agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only