4-Phenylcinnamic acid, 98%
4-Phenylcinnamic acid, 98%
4-Phenylcinnamic acid, 98%
Thermo Scientific Chemicals

4-Phenylcinnamic acid, 98%

CAS: 13026-23-8 | C15H12O2 | 224.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
50 g
Catalog number A12643.14
also known as A12643-14
Price (EUR)
236,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
236,00
Each
Chemical Identifiers
CAS13026-23-8
IUPAC Name(2Z)-3-{[1,1'-biphenyl]-4-yl}prop-2-enoic acid
Molecular FormulaC15H12O2
InChI KeyDMJDEZUEYXVYNO-FLIBITNWSA-N
SMILESOC(=O)C=C/C1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)224.0-230.0?C
Appearance (Color)White
Assay (Silylated GC)≥97.5%
4-Phenylcinnamic acid is used as pharmaceutical intermediates and for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenylcinnamic acid is used as pharmaceutical intermediates and for chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only