4,6-Dihydroxy-2-mercaptopyrimidine, 98%
4,6-Dihydroxy-2-mercaptopyrimidine, 98%
4,6-Dihydroxy-2-mercaptopyrimidine, 98%
Thermo Scientific Chemicals

4,6-Dihydroxy-2-mercaptopyrimidine, 98%

CAS: 504-17-6 | C4H4N2O2S | 144.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A12681.22
also known as A12681-22
Price (EUR)
118,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
118,00
Each
Chemical Identifiers
CAS504-17-6
IUPAC Name2-sulfanylidene-1,3-diazinane-4,6-dione
Molecular FormulaC4H4N2O2S
InChI KeyRVBUGGBMJDPOST-UHFFFAOYSA-N
SMILESO=C1CC(=O)NC(=S)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale pink
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
FormPowder
4,6-Dihydroxy-2-mercaptopyrimidine is used as a reagent in assaying malondialdehyde (theTBARS assay of lipid peroxidation). It is used to detect lipid oxidation, quantitating sialic acids and lipopolysaccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only