2,5-Dimethylaniline, 98+%
2,5-Dimethylaniline, 98+%
2,5-Dimethylaniline, 98+%
2,5-Dimethylaniline, 98+%
Thermo Scientific Chemicals

2,5-Dimethylaniline, 98+%

CAS: 95-78-3 | C8H11N | 121.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A12682.22
also known as A12682-22
Price (EUR)
39,36
Online Exclusive
46,30
Save 6,94 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
39,36
Online Exclusive
46,30
Save 6,94 (15%)
Each
Add to cart
Chemical Identifiers
CAS95-78-3
IUPAC Name2,5-dimethylaniline
Molecular FormulaC8H11N
InChI KeyVOWZNBNDMFLQGM-UHFFFAOYSA-N
SMILESCC1=CC=C(C)C(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless or pale yellow to orange to red/brown
Refractive Index1.5570-1.5610 @ 20?C
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥98.0%
2,5-Dimethylaniline is widely used as raw materials to produce imaging chemicals like organic dyes and pigments. It is also used in the production of antioxidants, pharmaceutical, agricultural, rubber chemicals and other target organic molecules. 2,5-Dimethylaniline was also used in the synthesis of nanocomposite of multi-walled carbon nanotubes embedded in poly(2,5-dimethylaniline). 2,5-Dimethylaniline was used to study the effect of metabolites formed from 2,5-xylidine by fungi on laccase activity. The Fourier transform infrared (FTIR) and Raman spectra of 2,5-dimethylaniline was studied5. It induced transcription of lccIV.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethylaniline is widely used as raw materials to produce imaging chemicals like organic dyes and pigments. It is also used in the production of antioxidants, pharmaceutical, agricultural, rubber chemicals and other target organic molecules. 2,5-Dimethylaniline was also used in the synthesis of nanocomposite of multi-walled carbon nanotubes embedded in poly(2,5-dimethylaniline). 2,5-Dimethylaniline was used to study the effect of metabolites formed from 2,5-xylidine by fungi on laccase activity. The Fourier transform infrared (FTIR) and Raman spectra of 2,5-dimethylaniline was studied5. It induced transcription of lccIV.

Solubility
Soluble in water <0.1 g/100 mL at 18°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. V Bavastrello.; E Stura.; S Carrara.; V Erokhin. Poly (2, 5-dimethylaniline)-MWNTs nanocomposite: a new material for conductometric acid vapors sensor. Sensors and Actuators B: Chemical. 2004, 98, (2-3), 247-253.
  2. EM Genies.; P Noel. Oxidation of 2, 5-dimethylaniline in NH 4 F· 2.35 HF medium Characteristics of the resulting conducting polymer. Journal of electroanalytical chemistry and interfacial electrochemistry. 1990, 296, (2), 473-490.