Sodium antimonyl L-tartrate, 98+%
Sodium antimonyl L-tartrate, 98+%
Sodium antimonyl L-tartrate, 98+%
Thermo Scientific Chemicals

Sodium antimonyl L-tartrate, 98+%

CAS: 34521-09-0 | C8H4Na2O12Sb2
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1000 g
Catalog number A12707.18
also known as A12707-18
Price (EUR)
29,30
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
29,30
Each
Chemical Identifiers
CAS34521-09-0
Molecular FormulaC8H4Na2O12Sb2
InChI KeyUZVPADYOEPQJSP-UHFFFAOYNA-J
SMILES[Na+].[Na+].[Sb+3].[Sb+3].[O-]C(C([O-])C([O-])=O)C([O-])=O.[O-]C(C([O-])C([O-])=O)C([O-])=O
Molecular Weight (g/mol)581.61
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Supplier's CofALoss on drying: 0 - 2 wt%
Appearance (Color)White
Assay (Titration)≥98%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only