3,5-Dibromobenzyl bromide, 99%
3,5-Dibromobenzyl bromide, 99%
3,5-Dibromobenzyl bromide, 99%
Thermo Scientific Chemicals

3,5-Dibromobenzyl bromide, 99%

CAS: 56908-88-4 | C7H5Br3 | 328.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A12726.14
also known as A12726-14
Price (EUR)
704,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
704,00
Each
Chemical Identifiers
CAS56908-88-4
IUPAC Name1,3-dibromo-5-(bromomethyl)benzene
Molecular FormulaC7H5Br3
InChI KeyPWTFRUXTAFBWBW-UHFFFAOYSA-N
SMILESBrCC1=CC(Br)=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)92.0-98.0?C
Appearance (Color)Pale yellow
Assay (GC)≥98.5%
3,5-Dibromobenzyl bromide, is an important raw material and intermediateintermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dibromobenzyl bromide, is an important raw material and intermediateintermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in toluene

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to water, oxidizing agents, bases, alcohols, amines, steel.
RUO – Research Use Only

General References:

  1. CP Yang.; TM Lee. Syntheses and properties of 4-hydroxy-3, 5-dibromobenzyl phosphonates and their flame-retarding effects on ABS copolymer.  Journal of Polymer Science Part A: Polymer. 1989 27, (7) , 2239-2251.
  2. R Epple.; M Azimioara.; R Russo.; B Bursulaya. 1, 3, 5-Trisubstituted aryls as highly selective PPARδ agonists. Bioorganic & Medicinal Chemistry Letters. 2006 16, (11) , 2969-2973.