5-Acetylthiophene-2-carbonitrile, 97%
5-Acetylthiophene-2-carbonitrile, 97%
5-Acetylthiophene-2-carbonitrile, 97%
5-Acetylthiophene-2-carbonitrile, 97%
Thermo Scientific Chemicals

5-Acetylthiophene-2-carbonitrile, 97%

CAS: 88653-55-8 | C7H5NOS | 151.18 g/mol
Have Questions?
Catalog NumberQuantity
A12765.88
also known as A12765-88
2.5 g
Catalog number A12765.88
also known as A12765-88
Price (EUR)
127,00
Each
Add to cart
Quantity:
2.5 g
Request bulk or custom format
Price (EUR)
127,00
Each
Add to cart
Specifications
Beilstein1306961
CAS88653-55-8
Chemical Name or Material5-Acetylthiophene-2-carbonitrile
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Formula Weight151.19
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only