Potassium sorbate, 99%
Potassium sorbate, 99%
Potassium sorbate, 99%
Thermo Scientific Chemicals

Potassium sorbate, 99%

CAS: 24634-61-5 | C6H7KO2 | 150.218 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A12844.30
also known as A12844-30
Price (EUR)
36,60
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
36,60
Each
Chemical Identifiers
CAS24634-61-5
IUPAC Namepotassium (2E,4E)-hexa-2,4-dienoate
Molecular FormulaC6H7KO2
InChI KeyCHHHXKFHOYLYRE-STWYSWDKSA-M
SMILES[K+].C\C=C\C=C\C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy MetalsMercury (Hg): ≤ 1ppm (non-U.S. specification)
Impurity contentArsenic (as As): ≤3ppm (non-U.S. specification)
Heavy Metals (as Pb)≤10ppm (non-U.S. specification)
Appearance (Color)White to pale cream
FormPowder or granules
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Potassium sorbate serves as an inhibitor for molds, yeasts in many food items like cheese, wine, yogurt, dried meats, apple cider, soft drinks, fruit drinks and baked goods. It is used in the production of syrup, milkshakes, creams, lotions, shampoos, makeup and sunscreen products. It is mainly used to increase the shelf life of various commercial products without affecting its taste, smell and color of the food. It is used as a preservative and considered to be a replacement for parabens.

Solubility
Soluble in water, alcohol, propylene glycol. Slightly soluble in acetoneand chloroform, corn oil and ether. Insoluble in benzene.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Hafner, S. D.; Windle, M.; Merrill, C.; Smith, M. L.; Franco, R. B.; Kung, L. Effects of potassium sorbate and Lactobacillus plantarum MTD1 on production of ethanol and other volatile organic compounds in corn silage. Anim. Feed Sci. Technol. 2015, 208, 79-85.
  2. Amirpour, M.; Arman, A.; Yolmeh, A.; Azam, M. A.; Sodium benzoate and potassium sorbate preservatives in food stuffs in Iran. Food Addit. Contam. B 2015, 8 (2), 142-148.