Guanidine carbonate, 98%
Guanidine carbonate, 98%
Guanidine carbonate, 98%
Thermo Scientific Chemicals

Guanidine carbonate, 98%

CAS: 593-85-1 | C3H12N6O3
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A12853.0B
also known as A12853-0B
Price (EUR)
58,70
Each
Quantity:
1000 g
Request bulk or custom format
Price (EUR)
58,70
Each
Chemical Identifiers
CAS593-85-1
IUPAC Namecarbonic acid; bis(guanidine)
Molecular FormulaC3H12N6O3
InChI KeySTIAPHVBRDNOAJ-UHFFFAOYSA-N
SMILESNC(N)=N.NC(N)=N.OC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
It is employed as a strong organic alkali, organic intermediate, in soap and cosmetic products, and in the textile industry. It is also used to make fine chemicals. It is used in various industries to solve different purpose such as detergents as a synergistic agent, pharmaceuticals as an antibiotic and microencapsulation process for carbonless paper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a strong organic alkali, organic intermediate, in soap and cosmetic products, and in the textile industry. It is also used to make fine chemicals. It is used in various industries to solve different purpose such as detergents as a synergistic agent, pharmaceuticals as an antibiotic and microencapsulation process for carbonless paper.

Solubility
Soluble in water 450 g/L (20°C); slightly soluble in alcohol and acetone.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. J.P. Varma. Inhibition of tobacco necrosis virus by guanidine carbonate.Virology.,1968,36(2), 305-308.
  2. Hugh M. Huffman.Thermal Data. XII. The Heats of Combustion of Urea and Guanidine Carbonate and their Standard Free Energies of Formation.J. Am. Chem. Soc.,1940,62(5), 1009-1011.