(+/-)-2-Chloropropionic acid, 94%
(+/-)-2-Chloropropionic acid, 94%
(+/-)-2-Chloropropionic acid, 94%
Thermo Scientific Chemicals

(+/-)-2-Chloropropionic acid, 94%

CAS: 598-78-7 | C3H5ClO2 | 108.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A12860.22
also known as A12860-22
Price (EUR)
28,90
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
28,90
Each
Chemical Identifiers
CAS598-78-7
IUPAC Name2-chloropropanoic acid
Molecular FormulaC3H5ClO2
InChI KeyGAWAYYRQGQZKCR-UHFFFAOYNA-N
SMILESCC(Cl)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥92.0%
Assay (Aqueous acid-base Titration)≥92.0 to ≤108.0%
Refractive Index1.4315-1.4385 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
RUO – Research Use Only