Bromotriphenylmethane, 98%
Bromotriphenylmethane, 98%
Bromotriphenylmethane, 98%
Bromotriphenylmethane, 98%
Thermo Scientific Chemicals

Bromotriphenylmethane, 98%

CAS: 596-43-0 | C19H15Br | 323.233 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A12864.18
also known as A12864-18
Price (EUR)
77,10
Precio exclusivo en nuestra web
90,70
Save 13,60 (15%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (EUR)
77,10
Precio exclusivo en nuestra web
90,70
Save 13,60 (15%)
Each
Add to cart
Chemical Identifiers
CAS596-43-0
IUPAC Name(bromodiphenylmethyl)benzene
Molecular FormulaC19H15Br
InChI KeyNZHXEWZGTQSYJM-UHFFFAOYSA-N
SMILESBrC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)151-156?C
FormPowder or crystalline powder
Appearance (Color)White to yellow
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
It is considered as a more reactive reagent than Chlorotriphenyl­methane, for the preparation of trityl esters. It is also used in one-pot conversion of amino acids to their N-trityl derivatives involving N,O-ditritylation and selective methanolysis of the trityl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is considered as a more reactive reagent than Chlorotriphenyl­methane, for the preparation of trityl esters. It is also used in one-pot conversion of amino acids to their N-trityl derivatives involving N,O-ditritylation and selective methanolysis of the trityl ester.

Solubility
Solubility in hot acetic acid is almost transparent.

Notes
Moisture sensitive. Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Alan C. Scott and John M. Tedder. The induced homolysis of bromotriphenylmethane by a nitroxide.J. Chem. Soc., Chem. Commun.,1979(2), 64b-65.
  2. A. E. Arbuzov and M. Sh. Bastonova. The tautomerism of carbostyril derivatives.Bulletin of the Academy of Sciences of the USSR, Division of chemical science,1952,1(5), 747-754.
  3. More reactive reagent than Chlorotriphenyl methane, A11799 for the preparation of trityl esters: J. Org. Chem., 27, 3595 (1962). One-pot conversion of amino acids to their N-trityl derivatives involves N,O-ditritylation and selective methanolysis of the trityl ester: Synthesis, 198 (1989). See also Appendix 6.