Dinonyl phthalate, mixture of isomers, 96%
Dinonyl phthalate, mixture of isomers, 96%
Dinonyl phthalate, mixture of isomers, 96%
Thermo Scientific Chemicals

Dinonyl phthalate, mixture of isomers, 96%

CAS: 84-76-4 | C26H42O4 | 418.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A12871.09
also known as A12871-09
Price (EUR)
40,70
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
40,70
Each
Chemical Identifiers
CAS84-76-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow-brown
FormViscous liquid
Refractive Index1.4820-1.4910 @ 20?C
Assay (GC)≥96.0% (sum of isomers)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only