4-Ethylpyridine, 98%
4-Ethylpyridine, 98%
4-Ethylpyridine, 98%
Thermo Scientific Chemicals

4-Ethylpyridine, 98%

CAS: 536-75-4 | C7H9N | 107.156 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A12883.18
also known as A12883-18
Price (EUR)
49,60
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
49,60
Each
Chemical Identifiers
CAS536-75-4
IUPAC Name4-ethylpyridine
Molecular FormulaC7H9N
InChI KeyVJXRKZJMGVSXPX-UHFFFAOYSA-N
SMILESCCC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4990-1.5030 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
4-Ethylpyridine is used to study the influence of counter-anion concentration on analyte retention.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethylpyridine is used to study the influence of counter-anion concentration on analyte retention.

Solubility
Slightly soluble in water (34 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Benjamin J. Coe.; Thomas J. Meyer.; Peter S. White. Control of axial ligand substitution in trans-bis(2,2'-bipyridine)ruthenium(II) complexes. Crystal and molecular structure of trans-(4-ethylpyridine)(dimethyl sulfoxide)bis(2,2'-bipyridine)ruthenium(II) hexafluorophosphate, trans-[Ru(bpy)2(4-Etpy)(DMSO)](PF6)2. Inorg. Chem. 1993, 32 (19), 4012-4020.
  2. S. C. Das.; P. Singh.; G. T. Hefter. The effects of 4-ethylpyridine and 2-cyanopyridine on zinc electrowinning from acidic sulfate solutions.J. Appl. Electrochem.1997, 27 (6), 738-744.