3,5-Dichloro-4-hydroxybenzoic acid, 97%
3,5-Dichloro-4-hydroxybenzoic acid, 97%
3,5-Dichloro-4-hydroxybenzoic acid, 97%
Thermo Scientific Chemicals

3,5-Dichloro-4-hydroxybenzoic acid, 97%

CAS: 3336-41-2 | C7H4Cl2O3 | 207.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12923.06
also known as A12923-06
Price (EUR)
32,20
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
32,20
Each
Chemical Identifiers
CAS3336-41-2
IUPAC Name3,5-dichloro-4-hydroxybenzoic acid
Molecular FormulaC7H4Cl2O3
InChI KeyAULKDLUOQCUNOK-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(Cl)=C(O)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)263.0-273.0?C
FormPowder
Assay (Silylated GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only