Allylbenzene, 98%
Allylbenzene, 98%
Allylbenzene, 98%
Thermo Scientific Chemicals

Allylbenzene, 98%

CAS: 300-57-2 | C9H10 | 118.179 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A12965.22
also known as A12965-22
Price (EUR)
180,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
180,00
Each
Chemical Identifiers
CAS300-57-2
IUPAC Name(prop-2-en-1-yl)benzene
Molecular FormulaC9H10
InChI KeyHJWLCRVIBGQPNF-UHFFFAOYSA-N
SMILESC=CCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.5090-1.5130 @ 20?C
FormLiquid
Allylbenzene is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allylbenzene is used in organic synthesis.

Solubility
Insoluble in water. Soluble in alcohol, ether and benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Shohei Suzuki.; Mitsuko Kato.; Shoichi Nakajima. Application of electrogenerated triphenylmethyl anion as a base for alkylation of arylacetic esters and arylacetonitriles and isomerization of allylbenzenes. Canadian Journal of Chemistry. 1994, 72 (2), 357-361.
  2. Doo-Jin Byun.; Sang Youl Kim . Selective Chain Transfer Reactions in Metallocene Catalyzed Copolymerization of Ethylene with Allylbenzene. Macromolecules. 2000, 33 (6), 1921-1923.