2-Iodoaniline, 98+%
2-Iodoaniline, 98+%
2-Iodoaniline, 98+%
Thermo Scientific Chemicals

2-Iodoaniline, 98+%

CAS: 615-43-0 | C6H6IN | 219.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
50 g
100 g
Catalog number A13059.18
Price (EUR)
195,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
195,00
Each
Chemical Identifiers
CAS615-43-0
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or needles
Identification (FTIR)Conforms
Assay (GC)≥98.0%
Appearance (Color)White to cream to orange to brown
2-Iodoaniline is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodoaniline is used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
It is sensitive to light. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Gozen Bereket; Evrim Hur; Yucel Sahin. Electrodeposition of polyaniline, poly(2-iodoaniline), and poly(aniline-co-2-iodoaniline) on steel surfaces and corrosion protection of steel. Applied Surface Science. 2005, 252, (5),1233-1244
  2. Qiuping Ding; Banpeng Cao; Xianjin Liu; Zhenzhen Zongand; Yi-Yuan Peng. Synthesis of 2-aminobenzothiazole via FeCl3-catalyzed tandem reaction of 2-iodoaniline with isothiocyanate in water. Green Chemistry. 2010, (9),1607-1610
  3. Quinolines may be synthesized by Pd-catalyzed reaction with allyl alcohols: Tetrahedron Lett., 32, 569 (1991).
  4. Reacts with unsymmetrical acetylenes in the presence of Palladium(II) acetate, 10516, to give indoles substituted at the 2- and/or 3-positions: J. Am. Chem. Soc., 113, 6689 (1991):
  5. Under similar conditions, ketones with an ɑ-methylene group also give indole derivatives: J. Org. Chem., 62, 2676 (1997).