Thermo Scientific Chemicals

4-Hydroxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: A13132.30
250 g, Each
Thermo Scientific Chemicals

4-Hydroxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: A13132.30
250 g, Each
Quantity
Catalog number: A13132.30
also known as A13132-30
Price (EUR)
Price: 338,00
Online price: 300,82
Your price:
Quantity
-

Chemical Identifiers

CAS
623-05-2
IUPAC Name
4-(hydroxymethyl)phenol
Molecular Formula
C7H8O2
InChI Key
BVJSUAQZOZWCKN-UHFFFAOYSA-N
SMILES
OCC1=CC=C(O)C=C1
Appearance (Color)
White to cream to pale yellow
Assay (Silylated GC)
≥98.5%
Identification (FTIR)
Conforms
Assay (HPLC)
≥98.5%
Melting Point (clear melt)
116.0-124.0°C

Description

4-Hydroxybenzyl alcohol is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzyl alcohol is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water (6.7 mg/ml at 20°C), dioxane (100 mg/ml), 1N NaOH (50 mg/ml), DMSO, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acid chlorides, acid anhydrides, bases, oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 - Harmful if swallowed

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing