3-Fluorobenzenesulfonyl chloride, 97%
3-Fluorobenzenesulfonyl chloride, 97%
3-Fluorobenzenesulfonyl chloride, 97%
Thermo Scientific Chemicals

3-Fluorobenzenesulfonyl chloride, 97%

CAS: 701-27-9 | C6H4ClFO2S | 194.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A13140.03
also known as A13140-03
Price (EUR)
29,70
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
29,70
Each
Chemical Identifiers
CAS701-27-9
IUPAC Name3-fluorobenzene-1-sulfonyl chloride
Molecular FormulaC6H4ClFO2S
InChI KeyOKYSUJVCDXZGKE-UHFFFAOYSA-N
SMILESFC1=CC(=CC=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to faintly turbid colorless to yellow to orange
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5275-1.5315 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only