2,4-Difluoro-1-nitrobenzene, 98%
2,4-Difluoro-1-nitrobenzene, 98%
2,4-Difluoro-1-nitrobenzene, 98%
2,4-Difluoro-1-nitrobenzene, 98%
Thermo Scientific Chemicals

2,4-Difluoro-1-nitrobenzene, 98%

CAS: 446-35-5 | C6H3F2NO2 | 159.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A13249.36
also known as A13249-36
Price (EUR)
308,00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (EUR)
308,00
Each
Add to cart
Chemical Identifiers
CAS446-35-5
IUPAC Name2,4-difluoro-1-nitrobenzene
Molecular FormulaC6H3F2NO2
InChI KeyRJXOVESYJFXCGI-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(F)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
Refractive Index1.5110-1.5150 @ 20°C
FormLiquid
Identification (FTIR)Conforms
2,4-Difluoronitrobenzene has been used in the synthesis of 2,4-difluoro-5-nitrobenzenesulfonic acid via sulfonation reaction, (±)-horsfiline and resin-bound-2-arylaminobenzimidazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoronitrobenzene has been used in the synthesis of 2,4-difluoro-5-nitrobenzenesulfonic acid via sulfonation reaction, (±)-horsfiline and resin-bound-2-arylaminobenzimidazoles.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
RUO – Research Use Only

General References:

  1. W. Cameron Black .; Benoit Guay.; Frank Scheuermeyer. Metalation of Nitroaromatics with in Situ Electrophiles. J. Org. Chem. 1997, 62 (3), 758-760 .
  2. Guanyu Yang.; Qiaohong Zhang .; Hong Miao.; Xinli Tong.; Jie Xu. Selective Organocatalytic Oxygenation of Hydrocarbons by Dioxygen Using Anthraquinones and N-Hydroxyphthalimide.Org. Lett. 2005, 7 (2),263-266 .
  3. Nucleophilic displacement of the activated fluorines with 2-ethyl-4-methylimidazole occurs predominantly at the 2-position, or, under more forcing conditions, at both 2- and 4-positions: J. Med. Chem., 34, 2671 (1991).