2-Benzoylbenzoic acid, 98%
2-Benzoylbenzoic acid, 98%
2-Benzoylbenzoic acid, 98%
Thermo Scientific Chemicals

2-Benzoylbenzoic acid, 98%

CAS: 85-52-9 | C14H10O3 | 226.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A13281.30
also known as A13281-30
Price (EUR)
89,65
Online exclusive
99,40
Save 9,75 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
89,65
Online exclusive
99,40
Save 9,75 (10%)
Each
Chemical Identifiers
CAS85-52-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale brown
FormPowder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Melting Point (clear melt)122-131°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only