trans,trans-2,4-Undecadienal, 90+%, remainder mainly trans,cis isomer
trans,trans-2,4-Undecadienal, 90+%, remainder mainly trans,cis isomer
trans,trans-2,4-Undecadienal, 90+%, remainder mainly trans,cis isomer
Thermo Scientific Chemicals

trans,trans-2,4-Undecadienal, 90+%, remainder mainly trans,cis isomer

CAS: 30361-29-6 | C11H18O | 166.264 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A13419.06
also known as A13419-06
Price (EUR)
23,60
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
23,60
Each
Chemical Identifiers
CAS30361-29-6
IUPAC Name(2E,4E)-undeca-2,4-dienal
Molecular FormulaC11H18O
InChI KeyUVIUIIFPIWRILL-XBLVEGMJSA-N
SMILESCCCCCCC=CC=CC=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5080-1.5160 @ 20?C
Assay (GC)>90.0% (major isomer)
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only