3-Bromo-5-chloro-2-hydroxybenzaldehyde, 97%
3-Bromo-5-chloro-2-hydroxybenzaldehyde, 97%
3-Bromo-5-chloro-2-hydroxybenzaldehyde, 97%
Thermo Scientific Chemicals

3-Bromo-5-chloro-2-hydroxybenzaldehyde, 97%

CAS: 19652-32-5 | C7H4BrClO2 | 235.461 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A13449.14
also known as A13449-14
Price (EUR)
161,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
161,00
Each
Chemical Identifiers
CAS19652-32-5
IUPAC Name3-bromo-5-chloro-2-hydroxybenzaldehyde
Molecular FormulaC7H4BrClO2
InChI KeyKNOYZLVIXXBBIB-UHFFFAOYSA-N
SMILESOC1=C(Br)C=C(Cl)C=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)81.5-90.5?C
Appearance (Color)Pale yellow to yellow
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only