4-Iodophenol, 98+%
4-Iodophenol, 98+%
4-Iodophenol, 98+%
Thermo Scientific Chemicals

4-Iodophenol, 98+%

CAS: 540-38-5 | C6H5IO | 220.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A13455.14
also known as A13455-14
Price (EUR)
93,80
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
93,80
Each
Chemical Identifiers
CAS540-38-5
IUPAC Name4-iodophenol
Molecular FormulaC6H5IO
InChI KeyVSMDINRNYYEDRN-UHFFFAOYSA-N
SMILESOC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)90.0-96.0?C
Appearance (Color)White to pale cream to cream to brown
Identification (FTIR)Conforms
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Iodophenol is a chemical used in chemiluminescence imaging assays and experiments, as diagnostic agents in order to detect cancer cells in patients. Also it is used in the synthesis of agonists towards the estrogen β receptor. Also used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display (LCD) chemicals.

Solubility
Soluble in alcohol and ether. Slightly soluble in water.

Notes
Light Sensitive, store in dark. Store in cool, dry place in tightly closed container. Store away from acid chlorides and acid anhydrides.
RUO – Research Use Only

General References:

  1. S J Brooks; E M Doyle; C Hewage; J P G Malthouse; W Duetz; K E O' Connor. Biotransformation of halophenols using crude cell extracts of Pseudomonas putida F6. Applied Microbiology and Biotechnology. 2004, 64(4), 486-492.
  2. A F Yakunin and P C Hallenbeck. A luminol/iodophenol chemiluminescent detection system for western immunoblots. Analytical Biochemistry, 1998, 258 (1), 146-149.