L-Phenylalanine ethyl ester hydrochloride, 98%
L-Phenylalanine ethyl ester hydrochloride, 98%
L-Phenylalanine ethyl ester hydrochloride, 98%
Thermo Scientific Chemicals

L-Phenylalanine ethyl ester hydrochloride, 98%

CAS: 3182-93-2 | C11H16ClNO2 | 229.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A13618.06
also known as A13618-06
Price (EUR)
37,90
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
37,90
Each
Chemical Identifiers
CAS3182-93-2
IUPAC Namehydrogen ethyl (2S)-2-amino-3-phenylpropanoate chloride
Molecular FormulaC11H16ClNO2
InChI KeyFPFQPLFYTKMCHN-PPHPATTJSA-N
SMILES[H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale blue
FormPowder or crystalline powder or crystals or needles
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Assay (Freebase GC)≥97.5%
Water Content (Karl Fischer Titration)≤1%
View more
L-Phenylalanine ethyl ester hydrochloride is used as an enzyme substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Phenylalanine ethyl ester hydrochloride is used as an enzyme substrate.

Solubility
Soluble in methanol and water.

Notes
Keep container tightly sealed. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature. Protect from humidity and water.
RUO – Research Use Only

General References:

  1. Martin J. O'Donnell.; Robin L. Polt. A mild and efficient route to Schiff base derivatives of amino acids. J. Org. Chem. 1982, 47 (132), 2663-2666.
  2. Yasuo Ariyoshi.; Tetsuo Yamatani.; Noboru Uchiyama.; Yohko Adachi.; Naotake Sato. The Synthesis of a Sweet Peptide, α-L-Aspartyl-L-phenylalanine Methyl Ester, without the Use of Protecting Groups. Bulletin of the Chemical Society of Japan. 1973, 46 (6), 1893-1895.