DL-Aspartic acid, 98+%
DL-Aspartic acid, 98+%
DL-Aspartic acid, 98+%
DL-Aspartic acid, 98+%
Thermo Scientific Chemicals

DL-Aspartic acid, 98+%

CAS: 617-45-8 | C4H7NO4 | 133.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A13646.36
also known as A13646-36
Price (EUR)
79,70
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (EUR)
79,70
Each
Add to cart
Chemical Identifiers
CAS617-45-8
IUPAC Name2-aminobutanedioic acid
Molecular FormulaC4H7NO4
InChI KeyCKLJMWTZIZZHCS-UHFFFAOYNA-N
SMILESNC(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
CommentSpecification differs for U.S. and non-U.S. material where indicated
FormCrystalline powder or powder or crystals
Identification (FTIR)Conforms (non-U.S. specification)
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
DL-Aspartic acid is used in studies on factors and conditions that enhance semen and sperm quality. Its conjugate base D-aspartate has potential use as a therapeutic agent in the treatment of schizophrenia-related symptoms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Aspartic acid is used in studies on factors and conditions that enhance semen and sperm quality. Its conjugate base D-aspartate has potential use as a therapeutic agent in the treatment of schizophrenia-related symptoms.

Solubility
Soluble in water, and hydrochloric acid. Insoluble in ether, and alcohol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. G. Macchia.; E. Topo.; N. Mangano.; E. D’Aniello.; R. Boni. dl-Aspartic acid administration improves semen quality in rabbit bucks. Animal Reproduction Science. 2010, 118 (2), 337-343.
  2. Susmita Singh.; Binod K Gogoi.; Rajib L Bezbaruah. Racemic resolution of some DL-amino acids using Aspergillus fumigatus L-amino acid oxidase. Current Microbiology. 2011, 63 (1), 94-99.