1,5-Hexadiene, 98%
1,5-Hexadiene, 98%
1,5-Hexadiene, 98%
Thermo Scientific Chemicals

1,5-Hexadiene, 98%

CAS: 592-42-7 | C6H10 | 82.146 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A13650.36
also known as A13650-36
Price (EUR)
426,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
426,00
Each
Chemical Identifiers
CAS592-42-7
IUPAC Namehexa-1,5-diene
Molecular FormulaC6H10
InChI KeyPYGSKMBEVAICCR-UHFFFAOYSA-N
SMILESC=CCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay from Supplier's CofA≥97.5%
1,5-Hexadiene is used to study the Pd(II)-catalyzed cycloisomerization of functionalized 1,5-hexadienes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Hexadiene is used to study the Pd(II)-catalyzed cycloisomerization of functionalized 1,5-hexadienes.

Solubility
Soluble in water(0.17g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Geoffrey W. Coates.; Robert M. Waymouth. Enantioselective cyclopolymerization of 1,5-hexadiene catalyzed by chiral zirconocenes: a novel strategy for the synthesis of optically active polymers with chirality in the main chain. J. Am. Chem. Soc. 1993, 115, (1), 91-98.
  2. Anne Lise Mogstad.; Robert M. Waymouth. Chain transfer to aluminum in the homogeneous cyclopolymerization of 1,5-hexadiene. Macromolecules. 1992, 25, (8), 2282-2284.