tert-Butyl bromide, 98+%, stab. with potassium carbonate
tert-Butyl bromide, 98+%, stab. with potassium carbonate
tert-Butyl bromide, 98+%, stab. with potassium carbonate
tert-Butyl bromide, 98+%, stab. with potassium carbonate
Thermo Scientific Chemicals

tert-Butyl bromide, 98+%, stab. with potassium carbonate

CAS: 507-19-7 | C4H9Br | 137.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A13727.22
also known as A13727-22
Price (EUR)
57,20
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
57,20
Each
Add to cart
Chemical Identifiers
CAS507-19-7
IUPAC Name2-bromo-2-methylpropane
Molecular FormulaC4H9Br
InChI KeyRKSOPLXZQNSWAS-UHFFFAOYSA-N
SMILESCC(C)(C)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to very pale yellow
Assay (GC)≥98.0%
Refractive Index1.4260-1.4300 @20?C
FormLiquid
tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.

Solubility
Immiscible with water.

Notes
Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ignaczak, A.; Schmickler, W. Mechanism and kinetics of electrochemical reduction of tert-butyl bromide molecule-improvement of theoretical model. Electrochim. Acta 2014, 134, 363-370.
  2. Ignaczak, A. Comparison of temperature effect on electroreduction of tert-butyl chloride and tert-butyl bromide -Theoretical study. Electrochim. Acta 2011, 56 (18), 6305-6311.