4-Allyloxybenzaldehyde, 97%
4-Allyloxybenzaldehyde, 97%
4-Allyloxybenzaldehyde, 97%
4-Allyloxybenzaldehyde, 97%
Thermo Scientific Chemicals

4-Allyloxybenzaldehyde, 97%

CAS: 40663-68-1 | C10H10O2 | 162.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A13773.09
also known as A13773-09
Price (EUR)
44,63
Online Exclusive
52,50
Save 7,87 (15%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (EUR)
44,63
Online Exclusive
52,50
Save 7,87 (15%)
Each
Add to cart
Chemical Identifiers
CAS40663-68-1
IUPAC Name4-(prop-2-en-1-yloxy)benzaldehyde
Molecular FormulaC10H10O2
InChI KeyTYNJQOJWNMZQFZ-UHFFFAOYSA-N
SMILESC=CCOC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.5660-1.5710 @ 20?C
FormLiquid
Free acid (titration)≤1.5%
Appearance (Color)Clear colorless to pale yellow
4-Allyloxybenzaldehyde is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Allyloxybenzaldehyde is used as pharmaceutical intermediates.

Solubility
Immiscible in water.

Notes
Use with adequate ventilation. Store under inert atmosphere. Protect from moisture and heat.
RUO – Research Use Only
Daisuke Nagai; Atsushi Sudo; Fumio Sanda; Takeshi Endo. Well-Defined Synthesis of Crosslinkable Polyester by Living Anionic Alternating Copolymerization of Ethylphenylketene with 4-Allyloxybenzaldehyde.Journal of Network Polymer,Japan.2001, 22 25-30.