Hexanoic acid is used to prepare esters by reacting with alcohols, which finds application in artificial flavors. It is also involved in the production of hexylphenols, hexanoates and caproates. Further, it is used as non-viral gene carrier as well as to protect tomato plants from Botrytis cinerea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hexanoic acid is used to prepare esters by reacting with alcohols, which finds application in artificial flavors. It is also involved in the production of hexylphenols, hexanoates and caproates. Further, it is used as non-viral gene carrier as well as to protect tomato plants from Botrytis cinerea.
Solubility
Miscible with diethyl ether and ethanol. Slightly miscible with water.
Notes
Store in a cool place. Incompatible with bases, oxidizing agents, reducing agents and allyl alcohol.
RUO – Research Use Only
General References:
- Ramesh, S.; Cherkupally, P.; Govender, T.; Kruger, H. G.; Albericio, F.; de la Torre, B. G. 6-(Bromomaleimido)hexanoic Acid as a Connector for the Construction of Multiple Branched Peptide Platforms. Org. Lett. 2015, 17 (3), 464-467.
- Lee, Y.; Choi, J. W.; Suh, D. J.; Ha, J. M.; Lee, C. H. Ketonization of hexanoic acid to diesel-blendable 6-undecanone on the stable zirconia aerogel catalyst. Appl. Catal., A 2015, 506, 288-293.