Tetraethylammonium bromide, 98%
Tetraethylammonium bromide, 98%
Tetraethylammonium bromide, 98%
Tetraethylammonium bromide, 98%
Thermo Scientific Chemicals

Tetraethylammonium bromide, 98%

CAS: 71-91-0 | C8H20BrN | 210.159 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A13835.0I
also known as A13835-0I
Price (EUR)
460,00
Each
Add to cart
Quantity:
5000 g
Request bulk or custom format
Price (EUR)
460,00
Each
Add to cart
Chemical Identifiers
CAS71-91-0
IUPAC Nametetraethylazanium bromide
Molecular FormulaC8H20BrN
InChI KeyHWCKGOZZJDHMNC-UHFFFAOYSA-M
SMILES[Br-].CC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
Loss on Drying≤2.0%
FormCrystals or powder or crystalline powder or lumps or fused solid
Appearance (Color)White to pale cream
Identification (FTIR)Conforms
Tetraethylammonium bromide is used as a phase transfer catalyst. It acts as a source of tetraethylammonium ions in pharmacological and physiological studies. It is also employed in organic chemical synthesis. It plays a major role in the preparation of tetraethylammonium superoxide from potassium superoxide for the conversion of primary alkyl halides to dialkyl peroxides. Furthermore, It is utilized as a catalyst in the oxidation of organic sulfide to sulfoxide using o-iodoxybenzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium bromide is used as a phase transfer catalyst. It acts as a source of tetraethylammonium ions in pharmacological and physiological studies. It is also employed in organic chemical synthesis. It plays a major role in the preparation of tetraethylammonium superoxide from potassium superoxide for the conversion of primary alkyl halides to dialkyl peroxides. Furthermore, It is utilized as a catalyst in the oxidation of organic sulfide to sulfoxide using o-iodoxybenzoic acid.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. Phase-transfer catalyst; see Appendix 2.
  2. For use in combination with anhydrous HBr as a reagent for selective monohydrobromination of an alkyne, see: Org. Synth., 76, 263 (1998).
  3. Takemoto, K.; Yamada, H. Development of rechargeable lithium-bromine batteries with lithium ion conducting solid electrolyte. J. Power Sources 2015, 281, 334-340.
  4. Abdel-Rahman, L. H.; El-Khatib, R. M.; Nassr, L. A.; Abu-Dief, A. M. Salts and Structural Effects on the base Catalyzed Hydrolysis of some Novel and Pharmacologically Active Iron(II) Azomethine Amino Acid Complexes. Int. J. Nano. Chem. 2015, 1 (1), 25-30.