3-Hydroxypyridine, 98%
3-Hydroxypyridine, 98%
3-Hydroxypyridine, 98%
Thermo Scientific Chemicals

3-Hydroxypyridine, 98%

CAS: 109-00-2 | C5H5NO | 95.101 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
Catalog number A13910.22
also known as A13910-22
Price (EUR)
63,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
63,00
Each
Chemical Identifiers
CAS109-00-2
IUPAC Namepyridin-3-ol
Molecular FormulaC5H5NO
InChI KeyGRFNBEZIAWKNCO-UHFFFAOYSA-N
SMILESOC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)123.0-130.0?C
Appearance (Color)Pale cream
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only