Suberic acid, 99%
Suberic acid, 99%
Suberic acid, 99%
Thermo Scientific Chemicals

Suberic acid, 99%

CAS: 505-48-6 | C8H14O4 | 174.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A13963.36
also known as A13963-36
Price (EUR)
111,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
111,00
Each
Chemical Identifiers
CAS505-48-6
IUPAC Nameoctanedioic acid
Molecular FormulaC8H14O4
InChI KeyTYFQFVWCELRYAO-UHFFFAOYSA-N
SMILESOC(=O)CCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (Silylated GC)≥98.5%
Identification (FTIR)Conforms
Melting Point (clear melt)139-146?C
View more
Suberic acid is used in drug synthesis and in the production of plastics. It is also used in the preparation of reduction-sensitive micelles, which affects their cellular uptake. Further, it is used in the delivery of anticancer drugs. In addition, it is used in the fluorescent detection of amidinium-carboxylate and amidinium formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suberic acid is used in drug synthesis and in the production of plastics. It is also used in the preparation of reduction-sensitive micelles, which affects their cellular uptake. Further, it is used in the delivery of anticancer drugs. In addition, it is used in the fluorescent detection of amidinium-carboxylate and amidinium formation.

Solubility
Soluble in water,dimethyl sulfoxide and methanol. Slightly soluble in ether. Insoluble in chloroform.

Notes
Incompatible with strong oxidizing agents, reducing agents and bases.
RUO – Research Use Only

General References:

  1. Cruickshank, D. L.; Younis, Y.; Njuguna, N. M.; Ongarora, D. S.; Chibale, K.; Caira, M. R. Alternative solid-state forms of a potent antimalarial aminopyridine: X-ray crystallographic, thermal and solubility aspects. CrystEngComm 2014, 16 (26), 5781-5792.
  2. Jiang, S.; Niu, S.; Zhao, Z. H.; Li, Z. J.; Li, Q. Synthesis of a series of novel heteroglycoclusters and homoglycoclusters and the study of their anti-adhesion activities. Carbohydr. Res. 2015, 414, 39-45.