2-Amino-4-nitrophenol, 98%, Thermo Scientific Chemicals
2-Amino-4-nitrophenol, 98%, Thermo Scientific Chemicals
2-Amino-4-nitrophenol, 98%, Thermo Scientific Chemicals
2-Amino-4-nitrophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-4-nitrophenol, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
A13964.14
also known as A13964-14
25 g
Catalog number A13964.14
also known as A13964-14
Price (EUR)
79,30
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
79,30
Each
Add to cart
Chemical Identifiers
CAS99-57-0
IUPAC Name2-amino-4-nitrophenol
Molecular FormulaC6H6N2O3
InChI KeyVLZVIIYRNMWPSN-UHFFFAOYSA-N
SMILESNC1=CC(=CC=C1O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow to orange to brown
FormPowder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only