p-Xylylene dichloride, 98%
p-Xylylene dichloride, 98%
p-Xylylene dichloride, 98%
Thermo Scientific Chemicals

p-Xylylene dichloride, 98%

CAS: 623-25-6 | C8H8Cl2 | 175.052 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A13976.14
also known as A13976-14
Price (EUR)
28,50
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
28,50
Each
Chemical Identifiers
CAS623-25-6
IUPAC Name1,4-bis(chloromethyl)benzene
Molecular FormulaC8H8Cl2
InChI KeyZZHIDJWUJRKHGX-UHFFFAOYSA-N
SMILESClCC1=CC=C(CCl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)98.0-104.0°C
p-Xylylene dichloride is a monomer used in preparation of poly(p-phenylene vinylene(PPV) nanotubes and nanorods by chemical vapor deposition (CVD) polymerization method. It was used in a study to investigate the effect of CVD monomer selection and reaction conditions on the resulting PPV polymer layer composition and luminescence properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Xylylene dichloride is a monomer used in preparation of poly(p-phenylene vinylene(PPV) nanotubes and nanorods by chemical vapor deposition (CVD) polymerization method. It was used in a study to investigate the effect of CVD monomer selection and reaction conditions on the resulting PPV polymer layer composition and luminescence properties.

Solubility
Insoluble in water

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases and active metals.
RUO – Research Use Only

General References:

  1. Kai Yu Wang; Qian Yang; Tai-Shung Chung; Raj Rajagopalan. Enhanced forward osmosis from chemically modified polybenzimidazole (PBI) nanofiltration hollow fiber membranes with a thin wall. Chemical Engineering Science. 2009, 64,(7), 1577-1584.
  2. M.P.Tsyurupa; V.A.Davankov. Hypercrosslinked polymers: basic principle of preparing the new class of polymeric materials. Reactive and Functional Polymers. 2002, 53,(2-3), 193-203.