Benzyltriphenylphosphonium chloride, 99%
Benzyltriphenylphosphonium chloride, 99%
Benzyltriphenylphosphonium chloride, 99%
Benzyltriphenylphosphonium chloride, 99%
Thermo Scientific Chemicals

Benzyltriphenylphosphonium chloride, 99%

CAS: 1100-88-5 | C25H22ClP | 388.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A14034.14
also known as A14034-14
Price (EUR)
34,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
34,00
Each
Add to cart
Chemical Identifiers
CAS1100-88-5
IUPAC Namebenzyltriphenylphosphanium chloride
Molecular FormulaC25H22ClP
InChI KeyUSFRYJRPHFMVBZ-UHFFFAOYSA-M
SMILES[Cl-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
Water Content (Karl Fischer Titration)≤0.5%
Melting Point>300?C
View more
Benzyltriphenylphosphonium chloride is used as the accelerator of the Bisphenol AF curing system for fluoroelastomer. Also used as phase-transfer catalysts in the synthesis of certain organic compounds. It is used as a reactant for the synthesis of chemical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyltriphenylphosphonium chloride is used as the accelerator of the Bisphenol AF curing system for fluoroelastomer. Also used as phase-transfer catalysts in the synthesis of certain organic compounds. It is used as a reactant for the synthesis of chemical compounds.

Solubility
Soluble in water and alcohol. Insoluble in acetone.

Notes
Hygroscopic. Store under inert gas. Store away from strong oxidizing agents, water/ moisture..
RUO – Research Use Only

General References:

  1. James McNulty; David McLeod. Amine- and sulfonamide-promoted Wittig olefination reactions in water. Chemistry: A European Journal. 2011, 17 (32), 8794-8798.
  2. G.I Mukhayer; S.S Davis. Interactions between large organic ions of opposite charge: VI. Coacervation in mixtures of sodium dodecyl sulfate and benzyltriphenylphosphonium chloride. Journal of Colloid and Interface Science. 1978, 66 (1), 110-117.
  3. Wittig olefination with carbonyl compounds can be carried out under basic, 2-phase conditions; the phosphonium salt acts as its own phase-transfer catalyst: Synthesis, 295 (1973). See also Appendix 1.
  4. Benzylidene triphenylphosphoranes (formed with n-BuLi) are coupled by reaction with sulfur to give stilbenes, with elimination of triphenylphosphine sulfide: Chem. Ber., 103, 2995 (1970).