2-(Trifluoromethyl)phenol, 98%
2-(Trifluoromethyl)phenol, 98%
2-(Trifluoromethyl)phenol, 98%
Thermo Scientific Chemicals

2-(Trifluoromethyl)phenol, 98%

CAS: 444-30-4 | C7H5F3O | 162.111 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A14231.06
also known as A14231-06
Price (EUR)
81,70
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
81,70
Each
Chemical Identifiers
CAS444-30-4
IUPAC Name2-(trifluoromethyl)phenol
Molecular FormulaC7H5F3O
InChI KeyZOQOPXVJANRGJZ-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder or fused solid and/or chunks
Assay (GC)≥97.5%
Water Content (Karl Fischer Titration)≤1.5%
Melting point37-47°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only