Bis(2-chloroethyl) ether, 99%
Bis(2-chloroethyl) ether, 99%
Bis(2-chloroethyl) ether, 99%
Thermo Scientific Chemicals

Bis(2-chloroethyl) ether, 99%

CAS: 111-44-4 | C4H8Cl2O | 143.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A14234.0B
also known as A14234-0B
Price (EUR)
215,00
Each
Quantity:
1000 g
Request bulk or custom format
Price (EUR)
215,00
Each
Chemical Identifiers
CAS111-44-4
IUPAC Name1-chloro-2-(2-chloroethoxy)ethane
Molecular FormulaC4H8Cl2O
InChI KeyZNSMNVMLTJELDZ-UHFFFAOYSA-N
SMILESClCCOCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.4555-1.4585 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Bis(2-chloroethyl) ether is used as a solvent and soil fumigant. It may be used in the preparation of macrocyclic polyethers, dibenzo-18-crown-6 polyether and dicyclohexyl-18-crown-6 polyether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-chloroethyl) ether is used as a solvent and soil fumigant. It may be used in the preparation of macrocyclic polyethers, dibenzo-18-crown-6 polyether and dicyclohexyl-18-crown-6 polyether.

Solubility
Soluble in water. (1.1 g/L) at 20°C.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
RUO – Research Use Only

General References:

  1. R D Lingg; W H Kaylor; S M Pyle; R G Tardiff. Thiodiglycolic acid: a major metabolite of bis(2-chloroethyl)ether. Toxicology and Applied Pharmacology.1979, 47 (1), 23-34.
  2. Alex Christensen; Mirat D. Gurol; Temesgen Garoma. Treatment of persistent organic compounds by integrated advanced oxidation processes and sequential batch reactor. Water Research.2009, 43 (16), 3910-3921.