Aniline, 99+%
Aniline, 99+%
Aniline, 99+%
Thermo Scientific™

Aniline, 99+%

CAS: 62-53-3 | C6H7N | 93.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1000 g
5000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A14443.22
also known as A14443-22
Price (EUR)
26,40
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
26,40
Each
Chemical Identifiers
CAS62-53-3
IUPAC Nameaniline
Molecular FormulaC6H7N
InChI KeyPAYRUJLWNCNPSJ-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.5845-1.5875 @ 20°C
Appearance (Color)Clear colorless to yellow or pale orange to orange to pale brown
FormLiquid
Assay (GC)≥99.0%
View more
Aniline is used as a solvent. It has been used as an antiknock compound for gasolines. It has a major application in the preparation of polyurethane which is used in making plastic, building thermal foam insulations and in refrigerators. It is widely used in the preparation of methylene dianiline and related compounds by reacting with formaldehyde. It combines with nitric acid and is utilized as rocket fuel for small missiles and jet assisted take-off. It acts as an additive to rubber and its derivatives such as phenylenediamines and diphenylamine are used as antioxidants. It finds application in dyes and pigments in agricultural and polymer industries. It is employed at a smaller scale in the production of the intrinsically conducting polymer polyaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aniline is used as a solvent. It has been used as an antiknock compound for gasolines. It has a major application in the preparation of polyurethane which is used in making plastic, building thermal foam insulations and in refrigerators. It is widely used in the preparation of methylene dianiline and related compounds by reacting with formaldehyde. It combines with nitric acid and is utilized as rocket fuel for small missiles and jet assisted take-off. It acts as an additive to rubber and its derivatives such as phenylenediamines and diphenylamine are used as antioxidants. It finds application in dyes and pigments in agricultural and polymer industries. It is employed at a smaller scale in the production of the intrinsically conducting polymer polyaniline.

Solubility
Miscible with water.

Notes
Light sensitive. Incompatible with oxidizing agents, iron and iron salts.
RUO – Research Use Only

General References:

  1. Qadir, M. I.; Scholten, J. D.; Dupont, J. Ionic liquid effect: selective aniline oxidative coupling to azoxybenzene by TiO2. Catal. Sci. Technol. 2015, 5 (3), 1459-1462.
  2. Fu, L.; Cai, W.; Wang, A.; Zheng, Y. Photocatalytic hydrogenation of nitrobenzene to aniline over tungsten oxide-silver nanowires. Mater. Lett. 2015, 142, 201-203.