Benzoylacetone, 98+%
Benzoylacetone, 98+%
Benzoylacetone, 98+%
Thermo Scientific Chemicals

Benzoylacetone, 98+%

CAS: 93-91-4 | C10H10O2 | 162.188 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A14537.30
also known as A14537-30
Price (EUR)
201,65
Online exclusive
224,00
Save 22,35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
201,65
Online exclusive
224,00
Save 22,35 (10%)
Each
Chemical Identifiers
CAS93-91-4
IUPAC Name1-phenylbutane-1,3-dione
Molecular FormulaC10H10O2
InChI KeyCVBUKMMMRLOKQR-UHFFFAOYSA-N
SMILESCC(=O)CC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream to pale yellow to pale brown
Melting Point (clear melt)53.0-60.0?C
Assay (GC)≥98.0%
Benzoylacetone has been used as a 1,3-dicarbonyl compound model for studying keto-enol equilibria in aqueous acid and micellar solutions. Additionally, it has been studied using gas-phase electron diffraction and quantum chemistry. It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoylacetone has been used as a 1,3-dicarbonyl compound model for studying keto-enol equilibria in aqueous acid and micellar solutions. Additionally, it has been studied using gas-phase electron diffraction and quantum chemistry. It is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Maxwell L.Eidinoff. Dissociation constants of acetylacetone, ethyl acetoacetate and benzoylacetone. J. Am. Chem. Soc. 1945, 67, (12),2072-2073
  2. Emilia Iglesias. Enolization of benzoylacetone in aqueous surfactant solutions: A novel method for determining enolization constants. J. Phys. Chem. 1996, 100, (30),12592-12599
  3. Dimetallated derivatives react with electrophiles preferentially at the terminal position; e.g. arylation by 2-chloroquinoline: J. Org. Chem., 37, 3199 (1972); J. Am. Chem. Soc., 97, 374 (1975). Reaction with aromatic esters gives 1,3-diaryl-1,3,5-pentanetriones, which can be cyclized with acid to -pyrones: Org. Synth. Coll., 5, 718, 721 (1973):