3-Bromoquinoline, 98%
3-Bromoquinoline, 98%
3-Bromoquinoline, 98%
Thermo Scientific Chemicals

3-Bromoquinoline, 98%

CAS: 5332-24-1 | C9H6BrN | 208.058 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
100 g
Catalog number A14573.09
also known as A14573-09
Price (EUR)
41,80
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
41,80
Each
Chemical Identifiers
CAS5332-24-1
IUPAC Name3-bromoquinoline
Molecular FormulaC9H6BrN
InChI KeyZGIKWINFUGEQEO-UHFFFAOYSA-N
SMILESBrC1=CN=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.6620-1.6680 @ 20°C
Appearance (Color)Clear colorless to yellow to orange
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only