4-Hydroxypyridine, 97%
4-Hydroxypyridine, 97%
4-Hydroxypyridine, 97%
Thermo Scientific Chemicals

4-Hydroxypyridine, 97%

CAS: 626-64-2 | C5H5NO | 95.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A14586.14
also known as A14586-14
Price (EUR)
48,50
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
48,50
Each
Chemical Identifiers
CAS626-64-2
IUPAC Name1,4-dihydropyridin-4-one
Molecular FormulaC5H5NO
InChI KeyGCNTZFIIOFTKIY-UHFFFAOYSA-N
SMILESO=C1C=CNC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)White to pale cream or pink to pale brown
Assay (Silylated GC)≥96.0%
Water Content (Karl Fischer Titration)≤1.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only