2-Acetylpyrrole, 98%
2-Acetylpyrrole, 98%
2-Acetylpyrrole, 98%
Thermo Scientific Chemicals

2-Acetylpyrrole, 98%

CAS: 1072-83-9 | C6H7NO | 109.128 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A14593.14
also known as A14593-14
Price (EUR)
160,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
160,00
Each
Chemical Identifiers
CAS1072-83-9
IUPAC Name1-(1H-pyrrol-2-yl)ethan-1-one
Molecular FormulaC6H7NO
InChI KeyIGJQUJNPMOYEJY-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)86.0-93.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only