2-Ethylaniline, 97%
2-Ethylaniline, 97%
2-Ethylaniline, 97%
Thermo Scientific Chemicals

2-Ethylaniline, 97%

CAS: 578-54-1 | C8H11N | 121.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A14984.22
also known as A14984-22
Price (EUR)
25,70
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
25,70
Each
Chemical Identifiers
CAS578-54-1
IUPAC Name2-ethylaniline
Molecular FormulaC8H11N
InChI KeyMLPVBIWIRCKMJV-UHFFFAOYSA-N
SMILESCCC1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5590-1.5620 @ 20?C
Appearance (Color)Clear colorless to yellow to red to red-brown or pink
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only