Methyl 3-nitrobenzoate, 98+%
Methyl 3-nitrobenzoate, 98+%
Methyl 3-nitrobenzoate, 98+%
Methyl 3-nitrobenzoate, 98+%
Thermo Scientific Chemicals

Methyl 3-nitrobenzoate, 98+%

CAS: 618-95-1 | C8H7NO4 | 181.147 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A15155.18
also known as A15155-18
Price (EUR)
39,20
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (EUR)
39,20
Each
Add to cart
Chemical Identifiers
CAS618-95-1
IUPAC Namemethyl 3-nitrobenzoate
Molecular FormulaC8H7NO4
InChI KeyAXLYJLKKPUICKV-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
Assay (GC)≥98.0%
FormCrystalline powder
Identification (FTIR)Conforms
Melting Point (clear melt)76-80?C
Methyl 3-nitrobenzoate was used in the preparation of iodoarene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-nitrobenzoate was used in the preparation of iodoarene.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Shawn J Stachel et. al. Conformationally biased P3 amide replacements of beta-secretase inhibitors. Bioorganic & Medicinal Chemistry Letters, 2006, 16(3), 641-644.
  2. L.-F. Jin; and F.-P. Xiao. Methyl 3-nitrobenzoate. Acta Cryst., 1963, 61o1237-o1238.